EMAIL THIS PAGE TO A FRIEND
Journal of the American Mosquito Control Association
Studies on the inheritance of repellent tolerances in Aedes aegypti.
PMID 8014634
Abstract
Two laboratory strains and 6 inbred strains of Aedes aegypti were tested against deet, ethyl hexanediol, dimethyl phthalate, and Indalone. Reciprocal crosses and backcrosses of 2 inbred strains were tested against deet only. Results obtained were compatible with a quantitative genetic model in which the effects of the factors involved were multiplicative. Certain inbred strains differed significantly from cognate laboratory and/or inbred strains in tolerance to one or more test materials. Heritability in the broad sense (H2) was estimated at 0.05 for deet, 0.22 for ethyl hexanediol, 0.48 for dimethyl phthalate, and 0.51 for Indalone. Partial dominance was observed in the inheritance of tolerance to deet.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E29125
2-Ethyl-1,3-hexanediol, 97%, Mixture of isomers
C8H18O2