European journal of biochemistry
C-type natriuretic peptide stimulates secretion of growth hormone from rat-pituitary-derived GH3 cells via a cyclic-GMP-mediated pathway.
PMID 8020502
Although C-type natriuretic peptide (CNP) has been shown to exist at the highest concentration in the anterior pituitary in rat tissues, its physiological role(s) there is (are) not clear. In this study, we report a novel function of CNP examined with anterior pituitary-derived cell lines, GH3 and AtT20/D16v-F2. Both CNP and atrial natriuretic peptide (ANP) increased cellular cGMP levels in both cell lines in dose-dependent manners. CNP, but not ANP, stimulated growth hormone (GH) release from GH3 cells. In contrast, neither ANP nor CNP had any significant effect on the corticotropin release from AtT20/D16v-F2 cells. An activator for cGMP-dependent protein kinase (cGK), dibutyryl cGMP, mimicked the stimulation of GH release from GH3 cells by CNP. Constitutive GH release from GH3 cells was greatly diminished in the presence of inhibitors for cAMP-dependent protein kinase, while stimulative GH release by CNP was not affected. However, inhibitors which can block cGK almost completely diminished the stimulative effect of CNP. An inhibitor for protein kinase C did not show any effect on either constitutive or CNP-stimulative GH release. Our observations indicate that the stimulation of GH release from GH3 cells by CNP is mediated mainly by the cGK signal-transduction pathway, not by cAMP-dependent protein kinase or protein kinase C, through a CNP-specific receptor (possibly ANP-B receptor). Thus, CNP may act as a local modulator in the anterior pituitary.
Related Materials
Product #
Molecular Formula
Add to Cart