EMAIL THIS PAGE TO A FRIEND
European journal of drug metabolism and pharmacokinetics
The metabolism of ethosuximide.
PMID 8020533
Abstract
The metabolism of the antiepileptic drug ethosuximide (3-ethyl-3-methylpyrollidine-2,5-dione) (I) in animals and humans is reviewed. Chiral aspects of the metabolism of ethosuximide are discussed. Clarification of the precise nature of the hydroxymetabolites of ethosuximide is presented.