EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Platinum (II) complexes of benzoic- and 3-methoxybenzoic acid hydrazides. Synthesis, characterization, and cytotoxic effect.
PMID 8027743
Abstract
The complexes [Pt(bah)2X2], [Pt(NH3)(bah)Cl2].0.5H2O, [Pt(mbah)2X2], and [Pt(NH3)(mbah)Cl2] (bah = benzoic acid hydrazide, mbah = 3-methoxybenzoic acid hydrazide; X = Cl, Br, I) have been prepared and characterized by elemental analysis, electric conductivity, IR, 1H NMR, and electronic spectra. A cis-square planar structure with hydrazide ligands coordinated via the NH2-groups has been proposed for these complexes. The complexes have shown a growth-inhibitory effect against Friend leukemia cells in culture comparable to that of the antitumor drug cisplatin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
117714
3-Methoxybenzoic acid, ReagentPlus®, 99%
C8H8O3
W394440
m-Anisic acid, ≥98%
C8H8O3