EMAIL THIS PAGE TO A FRIEND
Acta biochimica Polonica
The effects of captan and captafol on different bacterial strains and on c-mitosis in V79 Chinese hamster fibroblasts.
PMID 8030374
Abstract
The mutagenic activity of captan and captafol was tested using Ames strains and strains showing an SOS response. Captafol was mutagenic in S. typhimurium strain TA102 (uvr+) and captan in strain TA104 (uvrB). Both captan and captafol elicit damages in DNA recognized by correndonuclease II, as shown by the repair test, and induced the SOS repair system in E. coli PQ37 (uvrA) strain. Only captafol induced the SOS system in PQ35 (uvr+). The lack of induction of beta-galactosidase at nonpermissive temperature in E. coli MD332 (dnaCs uvrA) strain showed that neither chemical was able to produce DNA breaks. In V79 Chinese hamster fibroblasts higher induction of c-mitosis by captafol than by captan (22% and 15% over the control, respectively) was accompanied by a higher decrease in nonprotein sulfhydryl groups, mainly GSH (41% and 77%, respectively). The content of protein sulfhydryl groups was decreased by either fungicide to a similar extent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45365
Captafol, PESTANAL®, analytical standard
C10H9Cl4NO2S