EMAIL THIS PAGE TO A FRIEND
The American journal of emergency medicine
Cocaethylene: a current understanding of the active metabolite of cocaine and ethanol.
PMID 8031442
Abstract
Cocaine and ethanol abuse remains a major problem in our society. The active metabolite of the co-ingestion of cocaine and ethanol, cocaethylene, has been reported recently. This active and potentially toxic metabolite has enormous implications for emergency physicians. This article is a brief review of the current understanding of cocaethylene and its role as a serious toxin in the practice of emergency medicine.
Related Materials