EMAIL THIS PAGE TO A FRIEND
Journal of psychiatry & neuroscience : JPN
Re: Replacement of chlorpromazine with other neuroleptics: effect on abnormal skin pigmentation and ocular changes.
PMID 8031748
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F100
Fluspirilene
C29H31F2N3O