EMAIL THIS PAGE TO A FRIEND
Zhonghua nei ke za zhi
[Antidotal effect of sodium dimercapto-propane sulphonate in the treatment of acute poisoning of 2-dimethylamino-1,3-histhio-sulfopropanedisodium salt in 20 cases].
PMID 8033660