EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Lack of involvement of delta-1 opioid receptors in the development of physical dependence on morphine in mice.
PMID 8035333
Abstract
Previously, we have shown that the development of physical dependence on morphine in mice is inhibited substantially by treatment of mice with the highly selective, nonequilibrium delta-2 opioid receptor antagonist, naltrindole-5'-isothiocyanate. With the availability of the highly selective, nonequilibrium delta-1 opioid receptor antagonist, [D-Ala2,Leu5,Cys6]enkephalin, it was possible, in the present report, to examine the possible involvement of delta-1 opioid receptors in the development of opiate dependence. Mice were made physically dependent on morphine by s.c. implantation of morphine pellets (75-mg free base) for 3 days. The degree of dependence was quantified by determining the ED50 values of naloxone to precipitate withdrawal jumping and diarrhea. Neither sign of opiate withdrawal was affected by chronic treatment of animals with [D-Ala2,Leu5,Cys6]enkephalin during the morphine implant period. The data suggest that delta-1, as opposed to delta-2, opioid receptors are not involved in the development of physical dependence on morphine. This fact takes on added significance because the recently cloned delta opioid receptors appear to be the delta-2 subtype and the present data together with previous findings suggest that the cloned receptors may be proper models for the study of opiate dependence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N157
Naltrindole isothiocyanate hydrochloride, solid
C27H25N3O3S · HCl