EMAIL THIS PAGE TO A FRIEND
Anasthesiologie, Intensivmedizin, Notfallmedizin, Schmerztherapie : AINS
[Safety and tolerance of a new dipeptide-containing amino acid solution DP-Gln 20 versus a conventional amino acid solution Vamin 18EF in patients after elective colon surgery].
PMID 8043714
Abstract
Recent studies have demonstrated the important role of glutamine in postoperative metabolism. Using dipeptide-containing amino acid solutions it is possible to enhance glutamine supply. Safety and tolerance of DP-Gln 20 (gly-gln, gly-tyr) and Vamin 18EF were investigated in an open, prospective, randomised study. 16 patients received isonitrogenous parenteral nutrition over 4 days. There was no difference in clinical and biochemical patterns between both groups. Complications and adverse events due to the infused amino acid solutions were not observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3502
Gly-Tyr
C11H14N2O4