EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
Fc gamma-receptor activity of placental annexin II.
PMID 8047847
Abstract
We have previously produced a MoAb, B1D6, against a placental FcR. The antigen isolated using F(ab')2-fragments of B1D6 exhibits Fc-binding properties with low affinity for IgG. The antigen is a single-chained glycoprotein with a molecular weight of approximately 37 kDa and a pI of about 7.0-8.5. Amino acid sequences from enzymatic digests of the antigen indicated that it is annexin II. Immunoreactivity using anti-annexin antisera and purified placental annexin II have further established the specificity of B1D6 to annexin II. The B1D6 epitope appears to be intramembraneous and intracellular on placental syncytiotrophoblasts, monocytes and other cells investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
B1D6, 98042014, Reactivity: Anti-Annexin II, human