EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Respect for classics.
PMID 80558
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46908
Sulfisomidin, VETRANAL, analytical standard
C12H14N4O2S