EMAIL THIS PAGE TO A FRIEND
Journal of the American Oil Chemists' Society
Toxicology and biochemistry of butylated hydroxyanisole and butylated hydroxytoluene.
PMID 805808
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1082708
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
C15H24O