EMAIL THIS PAGE TO A FRIEND
Pflugers Archiv : European journal of physiology
Functional expression and properties of the human skeletal muscle sodium channel.
PMID 8058462
Abstract
Full-length deoxyribonucleic acid, complementary (cDNA) constructs encoding the alpha-subunit of the adult human skeletal muscle Na+ channel, hSkM1, were prepared. Functional expression was studied by electrophysiological recordings from cRNA-injected Xenopus oocytes and from transiently transfected tsA201 cells. The Na+ currents of hSkM1 had abnormally slow inactivation kinetics in oocytes, but relatively normal kinetics when expressed in the mammalian cell line. The inactivation kinetics of Na+ currents in oocytes, during a depolarization, were fitted by a weighted sum of two decaying exponentials. The time constant of the fast component was comparable to that of the single component observed in mammalian cells. The block of hSkM1 Na+ currents by the extracellular toxins tetrodotoxin (TTX) and mu-conotoxin (microCTX) was measured. The IC50 values were 25 nM (TTX) and 1.2 microM (microCTX) in oocytes. The potency of TTX is similar to that observed for the rat homolog rSkM1, but the potency of microCTX is 22-fold lower in hSkM1, primarily due to a higher rate of toxin dissociation in hSkM1. Single-channel recordings were obtained from outside-out patches of oocytes expressing hSkM1. The single-channel conductance, 24.9 pS, is similar to that observed for rSkM1 expressed in oocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
tsa201 Cell Line human, 96121229, embryonal kidney, SV40 transformed