EMAIL THIS PAGE TO A FRIEND
Annals of clinical biochemistry
Serum creatine kinase levels after a single intramuscular injection--dependence on injection volume.
PMID 8060101
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5520
Paraldehyde, ≥97%
C6H12O3