EMAIL THIS PAGE TO A FRIEND
Steroids
Synthesis of new 16-spirosteroids.
PMID 8073443
Abstract
Reaction of estrone methyl ether 1 with sodium hydride and 1,3-dibromopropane, 1,4-dibromobutane, and 1,5-dibromopentane, respectively, gives the 16-spirosteroid derivatives 2-4, which were reduced to the 17 beta-alcohols 7-9. Acetylation afforded the acetates 10-12. In the reaction of 1 and 1,3-dibromopropane a bis-allyl compound 5 and an annulated dihydropyran 6 were also formed. Cleavage of the ether moiety in 2 and 3 was accomplished with diisobutylaluminum hydride to give 3,17 beta-diols 13 and 14, respectively.
Related Materials