EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Determination of manganese peroxidase activity with 3-methyl-2-benzothiazolinone hydrazone and 3-(dimethylamino)benzoic acid.
PMID 8074299
Abstract
This method was proposed earlier for measuring glucose in a peroxidase-glucose oxidase system but has not been studied for determination of manganese peroxidase (MnP) activity. The assay is based on the oxidative coupling of 3-methyl-2-benzothiazolinone hydrazone (MBTH) and 3-(dimethylamino)benzoic acid (DMAB). The reaction of MBTH and DMAB in the presence of H2O2, Mn2+, and MnP gives a deep purple-blue color with a broad absorption band with a peak at 590 nm. The extinction coefficient is high (53,000 M-1 cm-1), so low MnP activities can be detected. Lignin peroxidase and laccase, usually present in cultures of white rot fungi, gave little or no interference at the concentrations tested. However, slight interference from very high LiP activity may occur at very low MnP activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0787
3-(Dimethylamino)benzoic acid, ≥97% (HPLC)
C9H11NO2