EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
A novel arginine (245) to glutamine change in exon 8 of the ornithine carbamoyl transferase gene in two unrelated children presenting with late onset deficiency and showing the same enzymatic pattern.
PMID 8081373