EMAIL THIS PAGE TO A FRIEND
Human mutation
Four newly identified ornithine transcarbamylase (OTC) mutations (D126G, R129H, I172M and W332X) in Japanese male patients with early-onset OTC deficiency.
PMID 8081398