EMAIL THIS PAGE TO A FRIEND
FEBS letters
Molecular characterization of a class IV human alcohol dehydrogenase gene (ADH7).
PMID 8082805
Abstract
Class IV alcohol dehydrogenase (ADH) is a form preferentially expressed in stomach. We report here the isolation and sequence determination of a novel human ADH gene (ADH7). Phylogenetic analysis strongly suggests that ADH7 is a functional class IV ADH gene.