EMAIL THIS PAGE TO A FRIEND
Medicinal research reviews
Potentiation of host defense mechanism against infection by a cytokine inducer, an acyl-MDP derivative, MDP-Lys(L18) (romurtide) in mice and humans.
PMID 8084203