EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Effects of cholecystokinin tetrapeptide and sulfated cholecystokinin octapeptide in rat models of anxiety.
PMID 8084519
Abstract
The effects of the acutely administered cholecystokinin (CCK) agonists CCK tetrapeptide (BOC-CCK-4) and sulfated CCK octapeptide (CCK-8S) were examined in four animal models of anxiety in rats. In the elevated plus maze, BOC-CCK-4 reduced the time spent in the open arms and the number of entries into the open arms. BOC-CCK-4 but not the anorectic acting CCK-8S increased the suppression of feeding in a conflict paradigm based on novelty suppressed feeding in hungry rats. In the two-compartment black-and-white box, BOC-CCK-4 decreased the time spent and locomotor activity in the white compartment. In the ultrasound vocalization test, using rat pups separated from the mother, BOC-CCK-4 increased the number of distress calls. No evidence was found for inducing anxiety-like behaviour by CCK-8S.