EMAIL THIS PAGE TO A FRIEND
The Journal of asthma : official journal of the Association for the Care of Asthma
The use of two inhaled beta agonists for the treatment of COPD: a summary of European data.
PMID 8096208
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32142
Pirbuterol acetate, VETRANAL, analytical standard
C12H20N2O3 · C2H4O2