EMAIL THIS PAGE TO A FRIEND
Human & experimental toxicology
Determination of oxidation phenotype by measurement of propafenone urinary metabolic ratios.
PMID 8096716
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC261
(±)-4-Hydroxydebrisoquin sulfate
C10H13N3O · 0.5H2SO4