EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Purified D-thyroxine in athyreotic patients.
PMID 8099168
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T34908
D-Thyroxine, 99%
C15H11I4NO4