EMAIL THIS PAGE TO A FRIEND
Oncogene
Induction of two alternatively spliced evi-1 proto-oncogene transcripts by cAMP in kidney cells.
PMID 8108138
Abstract
The evi-1 proto-oncogene is normally predominantly expressed in the kidney. We report here that evi-1 transcripts are also abundant in foetal kidney and expression is retained in primary kidney cell cultures. However available kidney cell lines express low or no evi-1 mRNA. In the human renal cell carcinoma cell line, A704, evi-1 is inducible approximately 16-fold by elevating intra-cellular cAMP levels with either forskolin or dibutyryl cAMP. TPA down-regulates evi-1 mRNA production and blocks forskolin mediated induction. Similar effects are seen in NIH3T3 cells and primary kidney cell cultures. Induction of evi-1 gene expression by forskolin does not alter the ratio of full length and an alternatively spliced transcript which encodes a protein lacking two repeats of the zinc finger motif. Potential regulation of evi-1 expression in kidney by hormones which modulate intra-cellular cAMP levels suggest that it can respond to environmental cues which might be important to the normal physiological role of this protein in kidney differentiation, development and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93020513
A.704