EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Selenol binds to iron in nitrogenase iron-molybdenum cofactor: an extended x-ray absorption fine structure study.
PMID 8108404
Abstract
The biological N2-fixation reaction is catalyzed by the enzyme nitrogenase. The metal cluster active site of this enzyme, the iron-molybdenum cofactor (FeMoco), can be studied either while bound within the MoFe protein component of nitrogenase or after it has been extracted into N-methylformamide. The two species are similar but not identical. For example, the addition of thiophenol or selenophenol to isolated FeMoco causes its rather broad S = 3/2 electron paramagnetic resonance signal to sharpen and more closely approach the signal exhibited by protein-bound FeMoco. The nature of this thiol/selenol binding site has been investigated by using Se-K edge extended x-ray absorption fine structure (EXAFS) to study selenophenol ligated to FeMoco, and the results are reported here. EXAFS data analysis at the ligand Se-K edge was performed with a set of software, GNXAS, that provides for direct calculation of the theoretical EXAFS signals and least-squares fits to the experimental data. Data analysis results show definitively that the selenol (and by inference thiol) binds to Fe at a distance of 2.4 A. In contrast, unacceptable fits are obtained with either Mo or S as the liganded atom (instead of Fe). These results provide quantitative details about an exchangeable thiol/selenol binding site on FeMoco in its isolated, solution state and establish an Fe atom as the site of this reaction. Furthermore, the utility of ligand-based EXAFS as a probe of coordination in polynuclear metal clusters is demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
375152
Benzeneselenol, 97%
C6H6Se