EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
One-step conversion of amino acids into N-menthyloxycarbonyl alkyl ester derivatives for chiral gas chromatography.
PMID 8109729
Abstract
Amino acids dissolved in aqueous methanol (or ethanol) and treated with optically active menthyl chloroformate were converted into N-menthyloxycarbonyl methyl (or ethyl) ester derivatives within a few minutes at room temperature. The obtained diastereomeric derivatives, with the exception of arginine and histidine, had suitable gas chromatographic properties allowing enantiomeric analysis of a large number of proteinogenic and synthetic amino acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245305
(1R)-(−)-Menthyl chloroformate, optical purity ee: 99% (GLC)
C11H19ClO2