EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong xi yi jie he za zhi Zhongguo Zhongxiyi jiehe zazhi = Chinese journal of integrated traditional and Western medicine
[Clinical study on cinobufotalin in treating chronic hepatitis B virus carriers].
PMID 8111200
Abstract
In a preliminary study, carriers of hepatitis B virus were treated with Cinobufotalin, a preparation from toad, for 1-3 course of treatment. Results indicated that negative conversion rate was markedly higher in the treated groups (21.09%, 38.89%, 63.64% and 50%) than that of control groups (2.1%, 10.87%, 33.33% and 7.7% P < 0.001). By the end of treatment, positive conversion rate of anti-HBs and anti-HBe were also markedly higher in the former (19.29% and 23.45%) than that in the latter (4.26% and 7.14% P < 0.001). During the follow-up of 3-6 months, the positive rate of HBeAg in treated group (50%) raised as compared with the control group (20% P = 0.08). The results indicated that Cinobufotalin had a significant effect in inhibiting the replication of HBV.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1147
Cinobufotalin
C26H34O7