EMAIL THIS PAGE TO A FRIEND
Journal of endodontics
Antibacterial effects of various endodontic medicaments on selected anaerobic bacteria.
PMID 8120484
Abstract
This study was undertaken to determine the antibacterial effects of various endodontic medicaments against six selected anaerobic bacteria. The experiment involved the testing of the vapors of six medicaments using agar plates streaked with the bacteria. A zone of inhibition was recorded for each plate and the results were analyzed statistically. Formocresol produced significantly larger zones of inhibition than any of the other medicaments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
531677
m-Tolyl acetate, 97%
C9H10O2