EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Alcoholysis of epsilon-decalactone with polyethylene glycol-modified lipase in 1,1,1-trichloroethane.
PMID 8123042
Abstract
Lipase from Pseudomonas cepacia was modified with 2,4-bis[O-methoxypoly(ethylene glycol)]-6-chloro-s-triazine, activated PEG2, to form PEG-lipase. The PEG-lipase is soluble and active in organic solvents. It catalyzes alcoholysis of racemic epsilon-decalactone with ethanol in 1,1,1-trichloroethane to form (R)-hydroxydecanoic acid ethyl ester. No alcoholysis of (S)-decalactone takes place. These results were discussed in relation to carbon number of n-alcohol, optimum temperature and comparison with modified and non-modified lipases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W361305
ε-Decalactone, ≥99%
C10H18O2