EMAIL THIS PAGE TO A FRIEND
Advances in enzymology and related areas of molecular biology
Lactose synthetase.
PMID 812340
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5507 Galactosyltransferase from bovine milk, lyophilized powder