EMAIL THIS PAGE TO A FRIEND
Steroids
N-alkylation of 17-azasteroids.
PMID 8140599
Abstract
N-Alkylation of 17-azasteroid lactams (16-oxo-17-azaandrost-5-en-3 beta-ol acetate 3 and its D-homo analog 4) was studied. It has been found that both lactams are readily alkylated with iodomethane or iodoethane. In contrast to this there was no reaction with 2-iodo-6-methylheptane due to the steric hindrance. 1,4-Addition of lactam to the conjugated systems was also studied. The addition to acrylonitrile proved to be easy compared to crotononitrile. However the efficient addition to the latter compound was also attained by using potassium t-butoxide as a base in t-butanol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122793
Allyl cyanide, 98%
C4H5N
252522
Crotononitrile, mixture of cis and trans, 99%
C4H5N