EMAIL THIS PAGE TO A FRIEND
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine
Structure of vulnibactin, a new polyamine-containing siderophore from Vibrio vulnificus.
PMID 8148612
Abstract
A new siderophore named vulnibactin has been isolated from low iron cultures of Vibrio vulnificus, a human pathogen. The structure was established as N-[3-(2,3-dihydroxybenzamido)propyl]-1,3-bis[2-(2-hydroxy-phenyl)- trans-5 - methyl-2-oxazoline-4-carboxamido]propane by a combination of acid hydrolysis, nuclear magnetic resonance spectroscopy and positive fast atom bombardment mass spectrometry. Vulnibactin is characterized as containing one residue of 2,3-dihydroxybenzoic acid as well as two residues of salicylic acid, both of which are involved in the formation of oxazoline rings with L-threonine bound to a norspermidine backbone. In addition, two other compounds with siderophore activity were purified and their structures were also determined. These two compounds provided further support for the structure of vulnibactin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
126209
2,3-Dihydroxybenzoic acid, 99%
C7H6O4