EMAIL THIS PAGE TO A FRIEND
Journal of applied toxicology : JAT
Oxadiazon.
PMID 8157873
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33382
Oxadiazon, PESTANAL®, analytical standard
C15H18Cl2N2O3