EMAIL THIS PAGE TO A FRIEND
Blood
Molecular abnormality of erythrocyte pyruvate kinase deficiency in the Amish.
PMID 8161798
Abstract
We describe the cellular and molecular biologic studies of the erythrocyte pyruvate kinase (PK) deficiency of the Amish deme in Pennsylvania. Nucleotide sequencing of the patient's PK gene showed a point mutation, CGC to CAC, corresponding to no. 1436 from the translational initiation site of the R-type PK (R-PK) mRNA, and it caused a single amino acid substitution from Arg to His at the 479th amino acid residue of the R-PK. The substituted Arg residue is located in the C domain of PK subunit, that is essential for both the intersubunit contact and the allosteric regulation. Because this enzyme shows the catalytic activity only as a dimer or tetramer, it is rational that the structural alteration would result in severe PK deficiency. To elucidate the effect of the PK deficiency on red blood cell (RBC) membrane, we performed the cellular studies of the patients' RBCs. Ouabain-insensitive K+ efflux was increased to 142% to 145% of normal controls and not inhibited by furosemide, as previously observed in HbSC disease RBCs.