EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Dipentafluorophenylcarbonate in the synthesis of oligonucleotides by the H-phosphonate method].
PMID 8166760
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
539899
Bis(pentafluorophenyl) carbonate, 97%
C13F10O3