EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Comparative cytotoxicity between butylated hydroxytoluene and its methylcarbamate derivative, terbucarb, on isolated rat hepatocytes.
PMID 8167443
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
246352
Methyl carbamate, 98%
C2H5NO2