EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Gas chromatography in the evaluation of 1,3-chlorobromopropane in the air of populated areas].
PMID 8168709
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9673
1-Bromo-3-chloropropane, for isolation of RNA
C3H6BrCl
B62404
1-Bromo-3-chloropropane, 99%
C3H6BrCl