EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Child and Adolescent Psychiatry
Update on lithium carbonate therapy in children and adolescents.
PMID 8169173
Abstract
The use of lithium to treat child and adolescent psychiatric disorders is becoming more common. Since the publication of the report of The Committee on Biological Aspects of Child Psychiatry of the American Academy of Child Psychiatry in 1978, a considerable body of literature has accumulated on the efficacy of lithium in treating adolescent bipolar disorders, childhood aggression, and behavioral disorders associated with mental retardation and developmental disorders. Efforts to understand lithium's mechanism(s) and refinements in psychiatric diagnosis have contributed to its growing use.