EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Effects of SH-group reagents on Ca2+ and K+ channel currents of pancreatic B-cells.
PMID 8179620
Abstract
The whole-cell patch-clamp technique has been applied to elucidate the effects of H2O2 and the SH-group reagent 2,2'-dithio-bis(5-nitropyridine) (DTBNP) on Ca2+ currents of mouse pancreatic B-cells. We also tested the effect of DTBNP on the whole-cell K+ ATP current. DTBNP (20 and 100 mumol/l) blocked both the current through Ca2+ channels (with Ba2+ as the charge carrier) and the K+ATP current in a concentration-dependent manner. The results suggest that the channels themselves or regulating proteins possess free SH-groups which may be of functional relevance. However, H2O2 did not influence the Ca2+ channel currents under standard whole-cell conditions as well as when recorded with the perforated patch technique. These results indicate that the two agents act differently on ion channel currents in B-cells although both are known to oxidize SH-groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158194
2,2′-Dithiobis(5-nitropyridine), 96%
C10H6N4O4S2