EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Direct evidence linking expression of matrix metalloproteinase 9 (92-kDa gelatinase/collagenase) to the metastatic phenotype in transformed rat embryo cells.
PMID 8183903
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CC1048 MMP-9, human, proform