EMAIL THIS PAGE TO A FRIEND
Bio-medical materials and engineering
A new determination method of network chain-length distribution of rubber vulcanizates by chemorheology.
PMID 8205061
Abstract
The structural changes of material caused by chemical reaction are reflected in the viscoelastic properties of these substances. Such a viscoelastic property induced by chemical reaction was termed chemorheology. Therefore, chemorheology is a relatively new discipline encompassing the physical and chemical aspects involved in the elucidation of the mechanisms, at a molecular level, of structural changes of polymers during their degradation. The studies presented here are the results stimulated further research in areas where this new determination will be more developed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329541
Dicumyl peroxide, 98%
C18H22O2