EMAIL THIS PAGE TO A FRIEND
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme
[Receptor agonists and antagonists: the present and future].
PMID 8210427
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05238
Erythromycin A enol ether, analytical standard
C37H65NO12