EMAIL THIS PAGE TO A FRIEND
Regulatory peptides
SR 48968, a neurokinin A (NK2) receptor antagonist.
PMID 8210494
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4267
Neurokinin A, ≥97% (HPLC)
C50H80N14O14S