EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Toxic injury to rat gut musculature following intraperitoneal administration of 2-t-butyl-4-methoxyphenol.
PMID 8223957
Abstract
The 100-fold increase in toxicity of intraperitoneal (i.p.) rather than orally administered 2-t-butyl-4-methoxyphenol (BHA) is adduced to the depressive effect which this compound exerts on the contractility of the gut musculature. A structure/activity relation study shows the t-butyl group on the benzene ring as being the major determinant of i.p. BHA toxicity. Contractile activity, elicited by field electrical stimulation, acetylcholine or Ba2+, of the ileum longitudinal muscle preparation from BHA-treated rats was greatly reduced 30 min after i.p. injection, and almost absent during the subsequent 48 h. Electron-microscope examination of ileum longitudinal muscle also showed partial destruction of cell membranes 4 h after BHA administration with subsequent mitochondrial swelling and destruction of cristae, myofibrillar fragmentation and cell necrosis. Comparable suppression of contractile activity and morphological damage were observed in BHA or t-butylbenzene incubated ileum segments where longitudinal smooth muscle contractility was irreversibly depressed in a time- and dose-dependent manner. These convergent findings point to the toxic effect of i.p. BHA on gut musculature with consequent impairment of intestinal transit.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19640
tert-Butylbenzene, analytical standard
C10H14
B90602
tert-Butylbenzene, 99%
C10H14