EMAIL THIS PAGE TO A FRIEND
IARC scientific publications
Formation of DNA adducts from 1-hydroxymethylpyrene in liver cells in vivo and in vitro.
PMID 8225483
Abstract
The binding of 1-hydroxymethylpyrene (HMP) and its active metabolites, 1-hydroxymethylpyrene sulfate (SMP) and 1-chloromethylpyrene (CMP), to DNA was studied. In the liver of rats, maximum adduct levels were observed 1.5 h after i.p. injection of HMP, followed by a relatively rapid decrease. Separate exposure of different liver cell types in vitro to HMP led to high adduct levels in parenchymal cells, intermediate levels (1/10) in endothelial cells and low levels (1/200) in Kupffer cells. The adduct patterns were similar in the different cell types. The same pattern was also obtained when isolated DNA was incubated with SMP or CMP. One of the four major spots co-chromatographed on TLC with a dG adduct, one with a dA adduct and one with both dG and dA adducts. The fourth spot did not co-chromatograph with any of the adducts observed in reactions with any nucleic acid homopolymer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
389439
1-Pyrenemethanol, 98%
C17H12O