EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Investigation of the usefulness of the photodynamic reaction for the purging of bone marrow from metastatic neuroblastoma cells.
PMID 8229464
Abstract
The neuroblastoma cell lines SK-N-LO, SK-PN-DW and IMR 5 were stained with Merocyanine 540 and exposed to white light. 99% of clonogenic tumour cells were destroyed while 60% of bone marrow progenitor cells survived this treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
SK-PN-DW, 94092301