EMAIL THIS PAGE TO A FRIEND
Stomatologiia
[Amben and galascorbin in the combined treatment of ulcerative-necrotic lesions of the oral mucosa].
PMID 8236292
Abstract
Clinical efficacy of ambene-galascorbic solution was studied in 58 patients with ulcerative necrotic lesions of the buccal mucosa. The results were assessed by clinical and laboratory parameters. Local application of ambene-galascorbic complex was found to be safe and effective in patients with ulcerative necrotic lesions of the buccal mucosa of various origin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
283746
4-(Aminomethyl)benzoic acid, 97%
C8H9NO2