EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effect of sublethal concentrations of glyphosate and dalapon on protein and aminotransferase activity in Pseudosuccinea columella.
PMID 8241626
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35562
Dalapon, PESTANAL®, analytical standard
C3H4Cl2O2